Learn More
IARS Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304747AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAS, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Isoleucine-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family and has been identified as a target of autoantibodies in the autoimmune disease polymyositis/dermatomyositis. Two alternatively spliced variants have been isolated that represent alternate 5′ UTRs.Specifications
IARS | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P41252 | |
IARS | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
IARS | |
Isoleucyl-tRNA synthetase, FLJ20736, IARS1, ILRS, PRO0785, IRS, ILERS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3376 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.