Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ iASPP, Mouse, Unlabeled, Clone: LX128.5,

Catalog No. MABC1166MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC1166MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1166MI Supplier MilliporeSigma™ Supplier No. MABC1166
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RelA-associated inhibitor (UniProt Q8WUF5; also known as Inhibitor of apoptosis-stimulating protein of p53, Inhibitor of ASPP protein, NF-kB-interacting protein 1, PPP1R13B-like protein, Protein iASPP) is encoded by the PPP1R13L (also known as IASPP, NKIP1, PPP1R13BL, RAI) gene (Gene ID 10848) in human. Inhibitor of apoptosis-stimulating protein of p53 (iASPP) belongs to the ASPP family of proteins (ASPP1, ASPP2, and iASPP) characterized by their ankyrin repeats (a.a. 659-691 and 692-724 in iASPP), SH3 domain (a.a. 758-820 in iASPP), and proline-rich (a.a. 54-602 in iASPP) sequences. iASPP is expressed at intercalated discs in post-mitotic cardiomyocytes, where it interacts with desmoplakin and desmin in cardiomyocytes to help maintain the integrity of desmosomes and intermediate filament networks. Desmosomes are anchoring junctions found in cells that endure physical stress such as cardiac myocytes or skin keratinocytes. Mutations of genes coding desmosome components, including IASPP/PPP1R13L, are found in human patients suffering from cardiomyopathy and in animal models of cardiomyopathy. iASPP deficiency caused by spontaneous mutation has been reported to be linked to a lethal autosomal recessive cardiomyopathy in Poll Hereford calves and Wa3 mice. iASSP-deficient mice with exon 8 deletion display features of arrhythmogenic right ventricular cardiomyopathy (ARVC) and die of sudden cardiac death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen iASPP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LX128.5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene PPP1R13L, IASPP, NKIP1, PPP1R13BL, RAI
Gene Accession No. Q8WUF5
Host Species Mouse
Immunogen Synthetic linear peptide corresponding to a sequence from the N-terminal region of human iASPP.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_006654
Test Specificity Clone LX128.5 immunostained intercalated discs in myocardial sections from wild-type, but not iASPP-deficient (iASPPδ8/δ8) adult mice (Notari, M., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(9):E973-E981).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.