Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ IB-MECA
GREENER_CHOICESDP

Catalog No. 10665 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-665 5 mg
10-662-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-665 Supplier R&D Systems™ Supplier No. 1066/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. A3 selective agonist

Potent and selective A3 adenosine receptor agonist (Ki values are 1.1, 54 and 56 nM for A3, A1 and A2A receptors respectively). Cardioprotective, reduces infarct size upon reperfusion in rats.

Chemical Identifiers

CAS 152918-18-8
Molecular Formula C18H19IN6O4

Specifications

Quantity 5 mg
Product Line Tocris
Formula Weight 510.29
Purity >97%
Molecular Weight (g/mol) 510.292
Chemical Name or Material IB-MECA
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.