Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Iba1/AIF1, Mouse, Alexa Fluor 647, Clone: 20A12.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABN92AF647
Description
Allograft inflammatory factor 1 (UniProt P55008; also known as AIF-1, Daintain, Interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, Protein G1) is encoded by the AIF1 (also known as G1, IBA1) gene (Gene ID 199) in human. Iba1/AIF1 is a 17kDa interferon-gamma- (IFN-gamma-) inducible, Ca(2+)-binding EF-hand protein. Iba1/AIF1 expression is mostly limited to the macrophage/monocyte lineage, and is augmented by cytokines, such as IFN-gamma. It is reported to be involved in neuronal function, inflammatory responses, allograft rejection, autoimmunity, and macrophage activation/function. It is also shown to be expressed in microglial cells in autominnume disesase models such as experimental autoimmune encephalomyelitis, neuritis and uveitis.
Specifications
| Iba1/AIF1 | |
| Monoclonal | |
| Please refer to lot specific datasheet. | |
| Purified mouse monoclonal IgG1κ antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % Sodium Azide. | |
| P55008 | |
| Linear peptide corresponding to human Iba1/AIF1. | |
| 100 μL | |
| Neuroscience | |
| NP_001614 | |
| Stable for 1 year at 2°-8°C from date of receipt. | |
| IgG1 κ |
| Immunofluorescence | |
| 20A12.1 | |
| Alexa Fluor 647 | |
| AIF1, G1, IBA1 | |
| Mouse | |
| Protein G Purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction