Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IBA57 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298047FR
Description
IBA57 Polyclonal specifically detects IBA57 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IBA57 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C1orf69, FLJ12734, FLJ13849, IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae), iron-sulfur cluster assembly factor for biotin synthase- and aconitase-like mitochondrial proteins, with a mass of 57kDa, mitochondrial, putative transferase C1orf69, mitochondrial | |
Produced in rabbits immunized with E. coli-derived Human IBA57 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
200205 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction