Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IBI Scientific™ Belly Bath
Shop All Stovall Life Science Inc. Products
Click to view available options
Electrical Requirements:
115 V 50/60 Hz
Description
Includes
Shaker/Bath combination comes with Belly Dancer Shaker modified to include temperature control plus IBI Scientific Hybridization Water Bath.
- Portable, versatile workhorse comprised of a lightweight, quick-heating water bath and The Belly Dancer™ Shaker
- Bath can be removed from shaker and used separately
- Microprocessor-controlled PID system
- Displays setpoint and actual water temperature
Shaker Features
- Gentle undulating motion
- Speed adjustable from 0 to 100rpm
- Orbital action with 1 in. (2.5cm) throw
- Platform pitch adjustable from flat to 8°
- 12 × 12 in. acrylic platform
- 5 lb. (2.25kg) load capacity
- Measures 15L x 15W x 9 in.H without bath on top; 15L x 15W x 17 in.H with bath lid
- Water Bath Features
- Temperature range of 30° to 75°C
- Temperature heat rate: approx. 3°C/min.
- Stability: ±0.2°C at setpoint
- PID microprocessor control maintains water temperature within ±0.1°C between 35° and 72°C
- Easily mobile: weighs just 7.5 lb. with lid (bath portion)
- Exterior measures 12.25L × 12.25W × 4 in.H (31 × 31 × 10.2cm)
- Stainless-steel chamber measures 10.38L × 10.38W × 3.25 in.D (26.4 × 26.4 × 8.3cm)
- Shaker and Water Bath together are perfect for Northern/Southern filter blot hybridizations, filter washes, in-situ washes and hybridizations, electrophoretic gel manipulation, acceleration of protein gel staining, hot staining protocols and recombinate DNA technology.
- Constant Temperature Bath used alone is ideal for protocols not requiring agitation such as heat inactivation of serums, restriction enzyme digests, and protein assays.
- Shaker is useful alone for non-temperature-sensitive stainings, washings and probes involved in gel and blotting techniques.
Specifications
Specifications
| Controller Type | Microprocessor |
| Temperature Range | 30°C to 75°C |
| Type | Belly Bath |
| Temperature Accuracy | ±0.1°C |
| Temperature Stability | ±0.2°C |
| Electrical Requirements | 115 V 50/60 Hz |
| Voltage | 115 V |
| Frequency | 50/60 Hz |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction