Learn More
Invitrogen™ iBlot™ 2 Transfer Stacks, PVDF, regular size
Description
iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. iBlot 2 Transfer Stacks are consumable stacks with an integrated pre-activated PVDF transfer membrane for dry blotting of proteins. Each iBlot 2 Transfer Stack contains a copper-coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins. This regular-size stack (13 x 8.3 cm) can be used with midi-size gels or two mini-size gels or E-PAGE 48 or 96 gels.
PVDF membrane
The PVDF membrane (0.2-μm pore size, low fluorescence) in this stack has higher binding capacity than nitrocellulose. It is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane due to hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.
Specifications
Specifications
| Quantity | 3 x 10 stacks |
| Format | Transfer Stack |
| Material | PVDF |
| Pore Size | 0.2 μm |
| Content And Storage | Store at room temperature. |
| Dimensions (LxW) | 13.5 x 8 cm |
| Product Line | iBlot |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.