Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IBRDC2 Antibody (OTI9H10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202128
Description
IBRDC2 Monoclonal specifically detects IBRDC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IBRDC2 | |
Monoclonal | |
0.8 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-100, Immunohistochemistry-Paraffin 1:50 | |
RNF144B | |
Human recombinant protein fragment corresponding to amino acids 1-256 of human RNF144B (NP_877434) produced in E. coli. | |
Affinity Purified | |
RUO | |
255488 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI9H10 | |
Unconjugated | |
bA528A10.3, E3 ubiquitin-protein ligase RNF144B, EC 6.3.2, EC 6.3.2.-, IBR domain containing 2, IBR domain-containing protein 2, IBRDC2, KIAA0161, p53-inducible RING finger protein, P53RFP, PIR2, ring finger protein 144BMGC71786 | |
Mouse | |
33.5 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction