Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ibritumomab Recombinant Mouse Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542307
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542307 Supplier Invitrogen™ Supplier No. MA542307
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Ibritumomab is a monoclonal antibody directed against the CD20 antigen found on pre-B and mature B lymphocytes (normal and malignant). Ibritumomab binding induces apoptosis in B lymphocytes in vitro. It is combined with the chelator tiuxetan, which acts as a specific chelation site for Yttrium-90 (Y-90). The monoclonal antibody acts as a delivery system to direct the radioactive isotope to the targeted cells, however, binding has been observed in lymphoid cells throughout the body and in lymphoid nodules in organs such as the large and small intestines. beta-emission induces cellular damage through the formation of free radicals (in both target cells and surrounding cells).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ibritumomab
Applications ELISA
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias MS4A-1
Host Species Mouse
Immunogen Human MS4A1(CD20,MS4A-1).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.