Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IBTK Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150033AF350
Description
IBTK Polyclonal antibody specifically detects IBTK in Human samples. It is validated for Western BlotSpecifications
IBTK | |
Polyclonal | |
Western Blot | |
Bruton agammaglobulinemia tyrosine kinase inhibitor, BTK-binding protein, BTKI, DKFZP564B116, IBtk, inhibitor of Bruton agammaglobulinemia tyrosine kinase, inhibitor of Bruton tyrosine kinase, inhibitor of Bruton's tyrosine kinase, inhibitor of Bruton's tyrosine kinase-alpha, inhibitor of Bruton's tyrosine kinase-beta, inhibitor of Bruton's tyrosine kinase-gamma, KIAA1417, MGC142256, MGC142258, RP1-93K22.1 | |
The immunogen for this product maps to a region between residue 1050 and 1100 of human Inhibitor of Bruton Agammaglobulinemia Tyrosine Kinase, using the numbering given in entry NP_056340.2 (GeneID 25998). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
25998 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction