Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IBTK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554895 Supplier Invitrogen™ Supplier No. PA554895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKEICEVALT EKLTLKNAAM LLEFAAMYSA KQLKLSCLQF IGLNMAALLE ARSLDVLSDG VLKDLSEFYR KMIPAMDRRV ITPYQDGPDI SYLEVEDGDI FLKEEINMEQ NHSETMFKKA K Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 79%.

The protein encoded by this gene binds to Bruton's tyrosine kinase (BTK) and downregulates BTK's kinase activity. In addition, the encoded protein disrupts BTK-mediated calcium mobilization and negatively regulates the activation of nuclear factor-kappa-B-driven transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IBTK
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IBTK
Gene Accession No. Q9P2D0
Gene Alias Bruton agammaglobulinemia tyrosine kinase inhibitor; BTBD26; BTK-binding protein; BTKI; IBTK; Inhibitor of Bruton tyrosine kinase; inhibitor of Bruton's tyrosine kinase-alpha; inhibitor of Bruton's tyrosine kinase-beta; inhibitor of Bruton's tyrosine kinase-gamma; KIAA1417; RP1-93K22.1
Gene Symbols IBTK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IBTK. Recombinant protein control fragment (Product #RP-102180).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.