Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBTK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590090 Supplier Thermo Scientific Supplier No. PA590090
Add to Cart
Add to Cart

IBTK Polyclonal Antibody

IBTK Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen IBTK
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IBTK
Gene Accession No. Q9P2D0
Gene Alias Bruton agammaglobulinemia tyrosine kinase inhibitor; BTBD26; BTK-binding protein; BTKI; IBTK; Inhibitor of Bruton tyrosine kinase; inhibitor of Bruton's tyrosine kinase-alpha; inhibitor of Bruton's tyrosine kinase-beta; inhibitor of Bruton's tyrosine kinase-gamma; KIAA1417; RP1-93K22.1
Gene Symbols IBTK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 900-1200 of human IBTK (NP_0563402).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25998
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.