Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ibudilast
GREENER_CHOICESDP

Catalog No. 169410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-941-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-941-0 Supplier R&D Systems™ Supplier No. 1694/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PDE inhibitor (non-selective)

Phosphodiesterase inhibitor (IC50 values are 53, 35, 48, 12 and 10 μM for PDE Ia, II, III, IV and V respectively). Inhibits platelet aggregation and is an orally-active cerebral vasodilator, bronchodilator and antiallergic agent.

Chemical Identifiers

CAS 50847-11-5
Molecular Formula C14H18N2O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 230.31
Purity >99%
Molecular Weight (g/mol) 230.311
Chemical Name or Material Ibudilast
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.