Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ibutilide hemifumarate
GREENER_CHOICESDP

Catalog No. 390850 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-085-0 Supplier R&D Systems™ Supplier No. 3908/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Blocks IKr, hERG and L-type Ca2+ channels; class III antiarrhythmic

Class III antiarrhythmic. Blocks the rapid component of delayed rectifier K+ current (IKr); also blocks hERG channels and L-type Ca2+ channels. Shown to activate a slow inward Na+ current in guinea pig ventricular cells. Used to terminate atrial fibrillation and atrial flutter; can prolong QT.

Chemical Identifiers

CAS 122647-32-9
Molecular Formula C44H76N4O10S2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 442.61
Purity >98%
Molecular Weight (g/mol) 885.23
Chemical Name or Material Ibutilide hemifumarate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.