Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICA1 Monoclonal Antibody (C7)
GREENER_CHOICE

Catalog No. PIMA541622
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541622 200 μL
PIMA543699 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541622 Supplier Invitrogen™ Supplier No. MA541622
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ICA1 Monoclonal Antibody for Western Blot

This gene encodes a protein with an arfaptin homology domain that is found both in the cytosol and as membrane-bound form on the Golgi complex and immature secretory granules. This protein is believed to be an autoantigen in insulin-dependent diabetes mellitus and primary Sjogren's syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICA1
Applications Western Blot
Classification Monoclonal
Clone C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ICA1
Gene Accession No. Q05084, Q63054
Gene Alias 69 kDa islet cell autoantigen; 69kDa; diabetes mellitus type I autoantigen; ica1; ica1 {ECO:0000312; ICA69; ICAp69; Islet cell autoantigen 1; islet cell autoantigen 1 isoform; islet cell autoantigen 1, 69 kDa; islet cell autoantigen 1, 69kDa; islet cell autoantigen p69; p69; RGD:621465}; testicular tissue protein Li 162; Unknown (protein for MGC:134001); zgc:92566
Gene Symbols ICA1
Host Species Mouse
Immunogen Recombinant protein Islet Cell Autoantigen 1. The antigen corresponds to amino acid range 1-258 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3382, 81024
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.