Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICA1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560005 Supplier Invitrogen™ Supplier No. PA560005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPSASLTSQE PSMGSEPLAH SSRFLPSQLF DLGFHVAGAF NNWVSQEESE LCLSHTDNQP VPSQSPKKLT RSPNNGNQD Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 63%.

ICA1L is a protein coding gene and gene ontology (GO) annotation includes acrosomal vesicle; protein domain specific binding; spermatid development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICA1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ICA1L
Gene Accession No. Q8NDH6
Gene Alias 1700030B17Rik; ALS2CR14; Als2cr15; amyotr; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 14; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 15; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 14 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein homolog; ICA1L; Ica69-related protein; islet cell autoantigen 1 like; islet cell autoantigen 1, 69kDa-like; islet cell autoantigen 1,69kDa-like; islet cell autoantigen 1-like; islet cell autoantigen 1-like protein
Gene Symbols ICA1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ICA1L. Recombinant protein control fragment (Product #RP-99665).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130026
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.