Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ICA1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598736
Description
ICA1L Polyclonal Antibody for Western Blot, ELISA
ICA1L is a protein coding gene and gene ontology (GO) annotation includes acrosomal vesicle; protein domain specific binding; spermatid development.
Specifications
ICA1L | |
Polyclonal | |
Unconjugated | |
ICA1L | |
1700030B17Rik; ALS2CR14; Als2cr15; amyotr; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 14; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 15; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 14 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein homolog; ICA1L; Ica69-related protein; islet cell autoantigen 1 like; islet cell autoantigen 1, 69kDa-like; islet cell autoantigen 1,69kDa-like; islet cell autoantigen 1-like; islet cell autoantigen 1-like protein | |
Rabbit | |
Protein G | |
RUO | |
130026 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8NDH6 | |
ICA1L | |
Recombinant Human Islet cell autoantigen 1-like protein (1-187AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction