Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-1/CD54 Antibody (112), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290847H
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Rat samples. It is validated for Sandwich ELISA Detection.Specifications
ICAM-1/CD54 | |
Monoclonal | |
HRP | |
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ICAM-1/CD54 (Q00238-1; Met1-Thr493). | |
0.1 mL | |
Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers | |
3383 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
112 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction