Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICAM-1 Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA129968
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129968 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129968 Supplier Invitrogen™ Supplier No. PA129968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-29968 detects ICAM1 from human, rhesus and cynomolgus monkey, baboon, an chimpanzee samples.

ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-1
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene ICAM1
Gene Accession No. P05362, Q28806
Gene Alias BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells
Gene Symbols ICAM1
Host Species Rabbit
Immunogen Full length protein (Human).
Purification Method IgG fraction
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101021731, 3383, 450196
Target Species Baboon, Chimpanzee, Human, Monkey
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.