Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2/CD102 Antibody (015), Janelia Fluor™ 646

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289421JF646
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
ICAM-2/CD102 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
15 | |
Sandwich ELISA Detection | |
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (NP_000864.2; Met1-Gln223). | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3384 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction