Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2/CD102 Antibody (041), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289424AF405
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot, ELISA.Specifications
ICAM-2/CD102 | |
Monoclonal | |
Alexa Fluor 405 | |
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (NP_777596.2; Met 1-Gln 223). | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3384 | |
Human | |
Purified |
Western Blot, ELISA | |
41 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CEACAM1/ CD66a Human ICAM1/ CD54 Human ICAM3/ CD50 Human CD31/ PECAM1 Mouse VCAM1/ L1CAM/ CD106 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction