Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2/CD102 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232236AF405
Description
ICAM-2/CD102 Polyclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Western BlotSpecifications
ICAM-2/CD102 | |
Polyclonal | |
Western Blot | |
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Intercellular Adhesion Molecule 2 using the numbering given in entry NP_001093256.1 (GeneID 3384). | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3384 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction