Learn More
ICAM-2/CD102 Antibody (B-T1), Alexa Fluor™ 750, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB10066624AF750
Catalog No. N1066624A70
Only null left
Add to Cart
Description
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ICAM-2/CD102
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
ICAM2
ICAM-2 transfected CHO cells
0.1 mL
Primary
Recognizes the human CD102 cell surface molecule, a 55-65kD glycoprotein also known as ICAM-2. NB100-66624 inhibits cell adhesion and T cell activation and also recognizes soluble ICAM-2.
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
B-T1
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Mouse
Protein A or G purified
Adaptive Immunity, Immunology
3384.0
Human
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.