Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2/CD102 Antibody (B-T1), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314603DL594
Description
ICAM-2/CD102 Monoclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
ICAM-2/CD102 | |
Monoclonal | |
DyLight 594 | |
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
ICAM-2 transfected CHO cell line | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3384 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA | |
B-T1 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction