Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2/CD102 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131612
Description
ICAM-2/CD102 Polyclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ICAM-2/CD102 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
ICAM2 | |
Recombinant protein encompassing a sequence within the center region of human ICAM2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
Rabbit | |
31 kDa | |
100 μL | |
Adaptive Immunity, Immunology | |
3384 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction