Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-2/CD102 Rabbit anti-Human, Clone: 33, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | ICAM-2/CD102 |
---|---|
Clone | 33 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot.Specifications
ICAM-2/CD102 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
Recombinant Human ICAM-2/CD102 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
33 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Immunology | |
0.2 um filtered solution in PBS | |
3384 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title