Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-3/CD50 Antibody (002), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289426R
Description
ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ICAM-3/CD50 | |
Monoclonal | |
DyLight 550 | |
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-3/CD50 (NP_002153.2; Met1-His485). | |
0.1 mL | |
Cellular Markers, Immunology | |
3385 | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Mouse ICAM1 Human ICAM Human VCAM1 Mouse VCAM1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction