Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-3/CD50 Antibody (CACT180A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260984
Description
ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Bovine, Water Buffalo samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
ICAM-3/CD50 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
Mouse | |
Unpurified | |
Cellular Markers, Immunology | |
3385 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
CACT180A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
ICAM3 | |
Bovine ConA stimulated mononuclear leukocytes | |
0.1 mg | |
Primary | |
Bovine, Water Buffalo | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction