Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-3/CD50 Antibody (ICAM 3.2), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250434PE
Description
ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Flow Cytometry.Specifications
ICAM-3/CD50 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
ICAM3 | |
ICAM-3/Fc chimeric fusion protein. | |
0.1 mL | |
Cellular Markers, Immunology | |
3385 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
ICAM 3.2 | |
Flow Cytometry | |
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction