Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-3/CD50 Antibody (OTI1E7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270975F
Description
ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human, Canine, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ICAM-3/CD50 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ICAM3 | |
Full length recombinant protein of human ICAM-3/CD50 (NP_002153) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Canine, Monkey | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1E7 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
Mouse | |
Affinity Purified | |
Cellular Markers, Immunology | |
3385.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction