Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-3/CD50 Antibody (SR1003), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | ICAM-3/CD50 |
---|---|
Clone | SR1003 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
ICAM-3/CD50 Monoclonal antibody specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
ICAM-3/CD50 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
A synthesized peptide derived from human ICAM-3/CD50 (Uniprot #: P32942) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1003 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cellular Markers, Immunology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3385 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title