Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-4 Polyclonal Antibody

ICAM-4 Polyclonal Antibody

Supplier:  Thermo Scientific PA550428

Catalog No. PA550428


Only null left
Add to Cart

Description

Description

ICAM-4 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ICAM-4
Polyclonal
Unconjugated
ICAM4
1810015M19Rik; CD242; CD242 antigen; ICAM4; ICAM-4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (Landsteiner-Wiener blood group); intercellular adhesion molecule 4 (LW blood group); intercellular adhesion molecule 4 isoform 3 anitgen LWa; intercellular adhesion molecule 4, Landsteiner-Wiener blood group; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW; LW blood group protein; LW glycoprotein
Rabbit
Antigen affinity chromatography
RUO
3386
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
2.1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q14773
ICAM4
Fusion protein corresponding to a region derived from internal residues of human intercellular adhesion molecule 4.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.