Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICAM-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577215
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577215 Supplier Invitrogen™ Supplier No. PA577215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE).

This gene encodes the Landsteiner-Wiener (LW) blood group antigen(s) that belongs to the immunoglobulin (Ig) superfamily, and that shares similarity with the intercellular adhesion molecule (ICAM) protein family. This ICAM protein contains 2 Ig-like C2-type domains and binds to the leukocyte adhesion LFA-1 protein. The molecular basis of the LW(A)/LW(B) blood group antigens is a single aa variation at position 100; Gln-100=LW(A) and Arg-100=LW(B). Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 50% glycerol, 1% BSA and 0.03% ProClin 300; pH 7.4
Gene ICAM4
Gene Accession No. Q14773, Q9ERM2
Gene Alias 1810015M19Rik; CD242; CD242 antigen; ICAM4; ICAM-4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (Landsteiner-Wiener blood group); intercellular adhesion molecule 4 (LW blood group); intercellular adhesion molecule 4 isoform 3 anitgen LWa; intercellular adhesion molecule 4, Landsteiner-Wiener blood group; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW; LW blood group protein; LW glycoprotein
Gene Symbols ICAM4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ICAM4: 85-180.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3386, 78369
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.