Learn More
Invitrogen™ ICAM-4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596676
Description
Immunogen sequence: VGSALGRRTK RAQSPKGSPL APSGTSVPFW VRMSPEFVAV QPGKSVQLNC SNSCPQPQNS SLRTPLRQGK TLRGPGWVSY QLLDVRAWSS LAHCLVTCAG KTRWATSRIT AYSVPGGLLG G; Positive Samples: HepG2, Jurkat, SW480, Mouse brain, Mouse spleen, Mouse heart, Mouse pancreas, Rat brain; Cellular Location: Cell membrane, Single-pass type I membrane protein.
This gene encodes the Landsteiner-Wiener (LW) blood group antigen(s) that belongs to the immunoglobulin (Ig) superfamily, and that shares similarity with the intercellular adhesion molecule (ICAM) protein family. This ICAM protein contains 2 Ig-like C2-type domains and binds to the leukocyte adhesion LFA-1 protein. The molecular basis of the LW(A)/LW(B) blood group antigens is a single aa variation at position 100; Gln-100=LW(A) and Arg-100=LW(B). Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
ICAM-4 | |
Polyclonal | |
Unconjugated | |
ICAM4 | |
1810015M19Rik; CD242; CD242 antigen; ICAM4; ICAM-4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (Landsteiner-Wiener blood group); intercellular adhesion molecule 4 (LW blood group); intercellular adhesion molecule 4 isoform 3 anitgen LWa; intercellular adhesion molecule 4, Landsteiner-Wiener blood group; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW; LW blood group protein; LW glycoprotein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
298702, 3386, 78369 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14773, Q9ERM2 | |
ICAM4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-140 of human ICAM4 (NP_0010342211). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.