Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICAM-5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547084
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547084 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547084 Supplier Invitrogen™ Supplier No. PA547084
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ICAM-5 and recombinant mouse (rm) ICAM-2 is observed, 5% cross-reactivity with recombinant rat ICAM-1, rhICAM-1, and rhICAM-3 is observed and less than 1% cross-reactivity with rmICAM-1 and rmVCAM-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Icam5
Gene Accession No. Q60625
Gene Alias CD50; Icam3; ICAM-3; Icam5; ICAM-5; intercellular adhesion molecule 3; intercellular adhesion molecule 5; intercellular adhesion molecule 5, telencephalin; telencephalin; Tlcn; TLN
Gene Symbols Icam5
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5 Leu31-Arg828 (Pro47Arg).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15898, 313785
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.