Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICAM-5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552632 Supplier Invitrogen™ Supplier No. PA552632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRSGELGAVI EGLLRVAREH AGTYRCEATN PRGSAAKNVA VTVEYGPRFE EPSCPSNWTW VEGSGRLFSC EVDGKPQPSV KCVGSGGATE GVLLPLAPPD PSPRAPRIPR VLAPGIYVCN ATNRHGSVAK TVVVS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Icam5
Gene Accession No. Q9UMF0
Gene Alias CD50; Icam3; ICAM-3; Icam5; ICAM-5; intercellular adhesion molecule 3; intercellular adhesion molecule 5; intercellular adhesion molecule 5, telencephalin; telencephalin; Tlcn; TLN
Gene Symbols Icam5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ICAM-5. Recombinant protein control fragment (Product #RP-90053).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.