Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM2 Mouse anti-Human, Clone:6B9G10, Abnova™

Catalog No. 89334299 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-299 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-299 Supplier Abnova Corporation Supplier No. MAB17988
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human ICAM2.

The protein encoded by this gene is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. This protein may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Specifications

Antigen ICAM2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6B9G10
Conjugate Unconjugated
Description intercellular adhesion molecule 2
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ICAM2
Gene Alias CD102
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 25-223 of human ICAM2.
Quantity 100 μg
Gene ID (Entrez) 3384
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.