Learn More
ICAM3 Mouse anti-Canine, Human, Clone: OTI1E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF507134

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ICAM3 (CD50) is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. This protein is constitutively and abundantly expressed by all leucocytes and may be the most important ligand for LFA-1 in the initiation of the immune response. It functions not only as an adhesion molecule, but also as a potent signaling molecule. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
ICAM3 | |
Monoclonal | |
Unconjugated | |
ICAM3 | |
CD50, CDW50, ICAM-R | |
Mouse | |
Affinity Chromatography | |
RUO | |
100683931, 3385 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P32942 | |
ICAM3 | |
Full length human recombit protein of human ICAM3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.