Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550387 Supplier Invitrogen™ Supplier No. PA550387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CTNNBIP1 protein.

The protein encoded by this gene binds CTNNB1 and prevents interaction between CTNNB1 and TCF family members. The encoded protein is a negative regulator of the Wnt signaling pathway. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CTNNBIP1
Gene Accession No. Q9NSA3
Gene Alias 1110008O09Rik; 2310001I19Rik; AW457332; beta-catenin-interacting protein 1; beta-catenin-interacting protein ICAT; catenin; catenin beta interacting protein 1; catenin, beta interacting protein 1; catenin, beta-interacting protein 1; Catnbip1; Ctnnbip1; ICAT; inhibitor of beta-catenin and Tcf-4
Gene Symbols CTNNBIP1
Host Species Rabbit
Immunogen Full length fusion protein of Human CTNNBIP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56998
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.