Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ICAT Recombinant Rabbit Monoclonal Antibody (8F7O5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542634
This item is not returnable.
View return policy
Description
Positive test controls include: TE-1. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MNREGAPGKS PEEMYIQQKV RVLLMLRKMG SNLTASEEEF LRTYAGVVNS QLSQLPPHSI DQGAEDVVMA FSRSETEDRR Q.
The protein encoded by this gene binds CTNNB1 and prevents interaction between CTNNB1 and TCF family members. The encoded protein is a negative regulator of the Wnt signaling pathway. Two transcript variants encoding the same protein have been found for this gene.
Specifications
ICAT | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NSA3 | |
CTNNBIP1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-81 of human CTNNBIP1 (Q9NSA3). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
8F7O5 | |
Unconjugated | |
CTNNBIP1 | |
1110008O09Rik; 2310001I19Rik; AW457332; beta-catenin-interacting protein 1; beta-catenin-interacting protein ICAT; catenin; catenin beta interacting protein 1; catenin, beta interacting protein 1; catenin, beta-interacting protein 1; Catnbip1; Ctnnbip1; ICAT; inhibitor of beta-catenin and Tcf-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56998 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction