Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICB-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598502
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598502 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598502 Supplier Invitrogen™ Supplier No. PA598502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ICB-1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

May constitute a control point in macrophage inflammatory response, promoting LPS-induced TNF production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICB-1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene THEMIS2
Gene Accession No. Q5TEJ8
Gene Alias basement membrane-induced; basement membrane-induced protein; C1orf38; Icb1; ICB-1; Induced by contact to basement membrane 1 protein; protein ICB-1; Protein THEMIS2; RGD1561016; THEMIS2; thymocyte selection associated family member 2; thymocyte-expressed molecule involved in selection protein 2
Gene Symbols THEMIS2
Host Species Rabbit
Immunogen Recombinant Human Protein THEMIS2 protein (62-177AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9473
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.