Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | ICB1 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ICB1 Polyclonal antibody specifically detects ICB1 in Human samples. It is validated for ImmunofluorescenceSpecifications
ICB1 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
basement membrane-induced, chromosome 1 open reading frame 38, ICB1, ICB-1, Induced by contact to basement membrane 1 protein, Protein ICB-1, protein THEMIS2, THEMIS2, Thymocyte-expressed molecule involved in selection protein 2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: GEREENPEFTSLAVGDRLEVLGPGQAHGAQGSDVDVLVCQRLSDQAGEDEEEECKEEAESPERVLLPFHFPGSF | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
9473 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title