Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICBP90 Binding Protein 1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282094C
This item is not returnable.
View return policy
Description
ICBP90 Binding Protein 1 Polyclonal antibody specifically detects ICBP90 Binding Protein 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ICBP90 Binding Protein 1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
C6orf107, dJ349A12.1, ICBP90, RP3-349A12.1, UHRF1 binding protein 1 | |
ICBP90 Binding Protein 1 antibody was raised against a 17 amino acid synthetic peptide near the center of human ICBP90 Binding Protein 1. The immunogen is located within amino acids 890 - 940 of ICBP90 Binding Protein 1. Amino Acid Squence: DSELSPSEDRELKSDAS | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
54887 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction