Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICDH Rabbit anti-E. coli, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS209025UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2090-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2090-25UL Supplier MilliporeSigma Supplier No. ABS209025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ICDH, Cat. No. ABS2090, is a rabbit polyclonal antibody that detects Isocitrate dehydrogenase [NADP]. It has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICDH
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Isocitrate dehydrogenase;ICDH (NADP);IDH
Gene Symbols ICDH
Host Species Rabbit
Immunogen His-tagged full length recombinant isocitrate dehydrogenase from Legionella pneumophila, strain Lp02.
Molecular Weight of Antigen 45 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This polyclonal antibody detects isocitrate dehydrogenase in E. coli and Legionella pneumophila.
Target Species E. coli
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.