Learn More
K1 Series Ice Cleat - Slim Profile
The K1 Slim is a one-size-fits-all traction solution designed for flat-soled footwear. The rotatable design combines a non-spiked anti-slip material with traditional ice-penetrating tungsten carbide studs to maximize safety and productivity when working indoors or outdoors.
Supplier: K1 Series V9770650O/S
This item is not returnable.
View return policy
Description
- Specially designed for flat-soled footwear found in walking and safety shoes
- Rotatable design provides increased traction for both indoor and outdoor use
- Non-spiked anti-slip material provides increased traction on wet surfaces
- Ice-penetrating tungsten carbide studs maximize grip on ice and snow
- Black or hi-viz adjustable strap delivers a perfect fit regardless of shoe size
- Ultra-thin rubber pad provides maximum comfort and is designed for all-day use
Specifications
Elastic | |
Outdoor Surfaces, Ice Surfaces | |
Universal | |
High Profile Tungston Carbide Molded in | |
Unisex |
Black | |
Non-Defined | |
Elastic | |
Reusable | |
Yes |