Learn More
CurranTaylor™ Ice Maker Installation Kit
SDPSCI_ED
Catalog No. S15531
Change view
Click to view available options
Inc. stainless steel flexible water line, various reducers, tees and fittings for drain assembly
Catalog No.
S15531 Inc. stainless steel flexible water line, various reducers, tees and fittings for drain assembly
1 options
Catalog No. S15531 Supplier CurranTaylor™ Supplier No. IMIKITBASE
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Designed for use with all major commercial ice maker machines
Kit Components:
  • Super value components
  • 152.4cm (60 in.) braided water connector (rated at 16.2L (4.3 gal./min.) at 50psi)
  • Lead-free in-line brass check valve
  • PVC adapters
  • Elbows and tees
Specifications
Product Type Installation Kit
For Use With (Application) Supplies the contents needed for initial installation
For Use With (Equipment) Designed for use with all major commercial ice maker machines
Includes Super value components, 60 in. braided water connector (rated at 4.3 gal./min. at 50psi), lead-free in-line brass check valve, PVC adapters, elbows and tees