Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566438 Supplier Invitrogen™ Supplier No. PA566438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKGSGTWEEKP KSHEAIQALN TWEVNKVTTS GLETFTATLR ESSATHSLVG EKHWTTASRS MSDRKRDILH ETKTQMEVRE MDKSVQTEKT IH Highest antigen sequence identity to the following orthologs - mouse 40%, rat 31%.

Component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III (PubMed:22195968, PubMed:23932780). Specifically acts as a scaffold protein that promotes the LEC complex formation and recruitment and RNA polymerase II occupancy at snRNA genes in subnuclear bodies (PubMed:23932780).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICE1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ICE1
Gene Accession No. Q9Y2F5
Gene Alias ATICE1; C77245; ICE1; INDUCER OF CBF EXPRESSION 1; interactor of little elongation complex ELL subunit 1; interactor of little elongator complex ELL subunit 1; KIAA0947; little elongation complex subunit 1; mKIAA0947; SCREAM; SCRM
Gene Symbols ICE1
Host Species Rabbit
Immunogen Recombinant Human ICE1. Recombinant protein control fragment (Product #RP-107113).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23379
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.