Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICE1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA562969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562969 Supplier Thermo Scientific Supplier No. PA562969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALSPELGASN FNDQKSSGIE YTKVVKGLTK IHSLPRSVFM KATKDGQCES QDPRIELTLN KPDFTSLIGS QAALIKSGLG FVKSTSWHHS DLLRKGG Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 46%.

ICE1 Polyclonal antibody specifically detects ICE1 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICE1
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ICE1
Gene Accession No. Q9Y2F5
Gene Alias C77245; ICE1; interactor of little elongation complex ELL subunit 1; interactor of little elongator complex ELL subunit 1; KIAA0947; little elongation complex subunit 1; mKIAA0947
Gene Symbols ICE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ICE1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23379
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.