Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICEBERG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558451
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558451 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558451 Supplier Invitrogen™ Supplier No. PA558451
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTVMDKARVL IDLVTGKGPK SCCKFIKHLC EEDPQLASKM GLH Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 49%.

Inhibits generation of IL-1-beta by interacting with caspase-1 and preventing its association with RIP2. Down-regulates the release of IL1B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICEBERG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CARD18
Gene Accession No. P57730
Gene Alias CARD18; caspase recruitment domain family member 18; caspase recruitment domain family, member 18; caspase recruitment domain-containing protein 18; Caspase-1 inhibitor Iceberg; ICEBERG; ICEBERG caspase-1 inhibitor; pseudo-ICE; UNQ5804; UNQ5804/PRO19611
Gene Symbols CARD18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ICEBERG. Recombinant protein control fragment (Product #RP-97330).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.