Learn More
BINDER™ ICH Light Module
SDP
Catalog No. p-200104333 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
KB PRO 720, KBF/KBF PRO 720
KB PRO/KBF/KBF PRO 260/470
ICH-Q1B light module
Module consists of:
  • VIS cassette
  • UVA cassette
  • Independent power supply
Specifications
For Use With (Equipment) KB PRO/KBF/KBF PRO 260/470
Material Glass
Type Lighting
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.